3-Bromocarbazole ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Bromo-9H-carbazole
B3458-5G
B3458-25G
TCIAB3458-5GEA
1100
NOK
TCIAB3458-5G
TCIAB3458-25G
3-Bromocarbazole ≥97.0% (by GC)
3-Bromocarbazole
Formel:
C₁₂H₈BrN MW: 246,11 g/mol Kokepunkt: 160°C |
MDL nummer:
MFCD00222621 CAS-nummer: 1592-95-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 197.0 to 202.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...