2-Bromo-1,3-dimethoxybenzene ≥98.0% (by GC)

Leverandør: TCI
Warning

B3488-5G B3488-1G B3488-25G
TCIAB3488-5GEA 460 NOK
TCIAB3488-5G TCIAB3488-1G TCIAB3488-25G
2-Bromo-1,3-dimethoxybenzene ≥98.0% (by GC)
2-Bromo-1,3-dimethoxybenzene
Formel: C₈H₉BrO₂
MW: 217,06 g/mol
Kokepunkt: 273°C
MDL nummer: MFCD07780177
CAS-nummer: 16932-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1b3e51055b795b93b97f9cd58a5046ea3ba8613&vid=e1b3e51055b795b93b97f9cd58a5046ea3ba8613&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR