2,3,6-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
T1563-1G
T1563-5G
TCIAT1563-1GEA
690
NOK
TCIAT1563-1G
TCIAT1563-5G
2,3,6-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2,3,6-Trimethoxybenzoic acid
Formel:
C₁₀H₁₂O₅ MW: 212,1992 g/mol |
MDL nummer:
MFCD00092587 CAS-nummer: 60241-74-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 148.0 to 151.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...