4-(2-Methoxyethyl)phenol ≥98.0% (by GC)
Leverandør: TCI
Warning
Synonymer: p-(2-Methoxyethyl) phenol
M1167-25G M1167-500G
TCIAM1167-25GEA 395 NOK
TCIAM1167-25G TCIAM1167-500G
4-(2-Methoxyethyl)phenol ≥98.0% (by GC)
4-(2-Methoxyethyl)phenol
Formel: C₉H₁₂O₂
MW: 152,19 g/mol
Kokepunkt: 125°C
MDL nummer: MFCD00017537
CAS-nummer: 56718-71-9
EINECS: 260-354-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73e25b9ec159f3425706caba698f66b3cc49224e&vid=73e25b9ec159f3425706caba698f66b3cc49224e&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C
Solubility in Methanol almost transparency