1-Bromo-3,5-di-tert-butylbenzene ≥98.0% (by GC)
Leverandør: TCI
B3547-25G
B3547-5G
TCIAB3547-25GEA
2000
NOK
TCIAB3547-25G
TCIAB3547-5G
1-Bromo-3,5-di-tert-butylbenzene ≥98.0% (by GC)
1-Bromo-3,5-di-tert-butylbenzene
Formel:
C₁₄H₂₁Br MW: 269,23 g/mol Kokepunkt: 158°C |
MDL nummer:
MFCD00796945 CAS-nummer: 22385-77-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...