Ethyl-2-methylnicotinate ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Ethyl 2-methylnicotinate

E0860-25G E0860-5G
TCIAE0860-25GEA 2450 NOK
TCIAE0860-25G TCIAE0860-5G
Ethyl-2-methylnicotinate ≥98.0% (by GC, titration analysis)
Ethyl-2-methylnicotinate
Formel: C₉H₁₁NO₂
MW: 165,19 g/mol
Kokepunkt: 125°C
MDL nummer: MFCD00006336
CAS-nummer: 1721-26-2
EINECS: 217-013-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1e878a8d702899b56c67915c19506b21c59a93a&vid=a1e878a8d702899b56c67915c19506b21c59a93a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR