4-Nitro-1H-pyrazole ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

N0862-25G N0862-5G
TCIAN0862-25GEA 2050 NOK
TCIAN0862-25G TCIAN0862-5G
4-Nitro-1H-pyrazole ≥98.0% (by GC, titration analysis)
4-Nitro-1H-pyrazole
Formel: C₃H₃N₃O₂
MW: 113,0800 g/mol
Kokepunkt: 323°C
MDL nummer: MFCD00159626
CAS-nummer: 2075-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db6341a96bcfdc99a388c2e3c94c510730c0a7c2&vid=db6341a96bcfdc99a388c2e3c94c510730c0a7c2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR