Benzhydrazide ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Benzoylhydrazine, Benzoic acid hydrazide, Benzohydrazide, Benzoic hydrazide

B0205-25G B0205-500G B0205-100G
TCIAB0205-25GEA 220 NOK
TCIAB0205-25G TCIAB0205-500G TCIAB0205-100G
Benzhydrazide ≥98.0% (by GC, titration analysis)
Benzhydrazide
Formel: C₇H₈N₂O
MW: 136,15 g/mol
Smeltepunkt: 112…114 °C
MDL nummer: MFCD00007596
CAS-nummer: 613-94-5
EINECS: 210-363-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=699b10187ac5ba011809798e5c274f97e55675ab&vid=699b10187ac5ba011809798e5c274f97e55675ab&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Bromination Method) min. 98.0 %
Melting point 112.0 to 116.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR