L(-)-Cystin ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Synonymer: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
C0519-25G C0519-500G
TCIAC0519-25GEA 260 NOK
TCIAC0519-25G TCIAC0519-500G
L(-)-Cystin ≥98.0% (by HPLC, total nitrogen)
L(-)-Cystin
Formel: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240,3 g/mol
Kokepunkt: 428 °C (1013 hPa)
Smeltepunkt: 261…262 °C
Tetthet: 1,677 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064228
CAS-nummer: 56-89-3
EINECS: 200-296-3
Merck Indeks: 14,02782
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8744d69264eb6e03d27a0d563aa6cfe1871492cd&vid=8744d69264eb6e03d27a0d563aa6cfe1871492cd&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -215 to -230 deg (C=1, 1mol/L HCl)
Solubility in HCl transparency