3-(Hydroxymethyl)indole ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: 3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol

I0496-25G I0496-5G
TCIAI0496-25GEA 1900 NOK
TCIAI0496-25G TCIAI0496-5G
3-(Hydroxymethyl)indole ≥98.0% (by HPLC, total nitrogen)
3-(Hydroxymethyl)indole
Formel: C₉H₉NO
MW: 147,18 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00005632
CAS-nummer: 700-06-1
EINECS: 211-836-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32adee6338a022600954052326beb972e80b8139&vid=32adee6338a022600954052326beb972e80b8139&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR