2-Amino-5-nitro-6-picoline ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
6-Amino-2-methyl-3-nitropyridine, 2-Amino-5-nitro-6-methylpyridine, 2-Amino-6-methyl-5-nitropyridine, 6-Amino-3-nitro-2-picoline, 6-Methyl-5-nitro-2-pyridylamine
A2326-1G
A2326-5G
TCIAA2326-1GEA
405
NOK
TCIAA2326-1G
TCIAA2326-5G
2-Amino-5-nitro-6-picoline ≥98.0% (by GC)
2-Amino-5-nitro-6-picoline
Formel:
C₆H₇N₃O₂ MW: 153,14 g/mol |
MDL nummer:
MFCD00053582 CAS-nummer: 22280-62-2 |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 189.0 to 193.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...