1H,1H,2H-Perfluor-1-deken ≥97.0% (by GC)

Leverandør: TCI
Warning

H0846-25G H0846-5G
TCIAH0846-25GEA 1150 NOK
TCIAH0846-25G TCIAH0846-5G
1H,1H,2H-Perfluor-1-deken ≥97.0% (by GC)
1H,1H,2H-Perfluor-1-deken
Formel: C₁₀H₃F₁₇
MW: 446,11 g/mol
Kokepunkt: 147°C
Tetthet: 1,65 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00039246
CAS-nummer: 21652-58-4
EINECS: 244-503-5
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cf60d6beaa5cb86d4050cc12b0af6d968600a58&vid=7b574e47a0db9f7ba24953b68ae273bed305785a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR