1,2,4-Trifluoro-5-nitrobenzene ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
2,4,5-Trifluoronitrobenzene
T1589-25G
T1589-5G
TCIAT1589-25GEA
715
NOK
TCIAT1589-25G
TCIAT1589-5G
1,2,4-Trifluoro-5-nitrobenzene ≥98.0% (by GC)
1,2,4-Trifluoro-5-nitrobenzene
Formel:
C₆H₂F₃NO₂ MW: 177,08 g/mol Kokepunkt: 195°C Smeltepunkt: –11 °C Tetthet: 1,54 g/cm³ (20 °C) Flash punkt: 89°C |
MDL nummer:
MFCD00007051 CAS-nummer: 2105-61-5 EINECS: 218-281-5 FN: 2810 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown to Dark green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...