Screw neck vials, N 8
Leverandør: MACHEREY-NAGEL
70213 70213.2 702860 702893 702004
218-1105EA 385 NOK
218-1105 MANA70213.2 MANA702860 MANA702893 MANA702004
Screw neck vials, N 8
Vialer
These screw neck vials are characterised by a small opening and 8-425 thread.
  • Made of clear and amber glass
  • Available with flat or conical bottom
  • Different inserts available
Often used on instruments of Dionex (Thermo Scientific), Knauer, Thermo Scientific, Varian (Agilent), VWR® (Merck / Hitachi) and others.
Vials with closed top screw closures are suitable for sample storage.
The suitable screw closures are ready assembled.
- Septa made from economical Red Rubber/FEP, high quality silicone/PTFE or from virgin PTFE
- Silicone/PTFE septa areMore Product Information