Analytical balances, Practum
Leverandør: Sartorius Balances
PRACTUM224-1S PRACTUM124-1S PRACTUM64-1S
611-2660EA 36100 NOK
611-2660 611-2657 611-2889
Analytical balances, Practum
Vekter
A practical basic balance that incorporates Sartorius' renowned weighing technology to deliver reliable professional results. Weighing applications include general weighing, density, percentage, check weighing, peak hold, counting and averaging. Results can be transferred via a mini-USB interface to a printer or a computer directly into a Microsoft Excel® spreadsheet without additional software. Menu with choice of 10 languages.
  • High contrast colour touch screen
  • Automatic calibration and adjustment possible with an optional external weight
  • Sturdy draught shield can be completely disassembled for easy cleaning
  • Supervisor lock to prevent any unintentional changes to settings
  • Built-in overload protection
Leveringsinformasjon: Supplied with universal power adapter, in-use cover and application guide but without calibration weight.