Lokk, Skrulokk for skruhals hetteglass, N 9
Leverandør: MACHEREY-NAGEL
702286 702287 702288 702288.1 702287.1 702030 702035 702034 702028 702027 702732 702032 702039 702033 702038 702037 702081 702026 702029 702031 702036 702040 702080 702082 702083 702084 702085 702107 702109 702147 702155 702156 702158 702159 702160 702161 702162 702163 702164 702165 702402
MANA702286EA 745 NOK
MANA702286 MANA702287 MANA702288 MANA702288.1 MANA702287.1 MANA702030 MANA702035 MANA702034 MANA702028 MANA702027 MANA702732 MANA702032 MANA702039 MANA702033 MANA702038 MANA702037 MANA702081 MANA702026 MANA702029 MANA702031 MANA702036 MANA702040 MANA702080 MANA702082 MANA702083 MANA702084 MANA702085 MANA702107 MANA702109 MANA702147 MANA702155 MANA702156 MANA702158 MANA702159 MANA702160 MANA702161 MANA702162 MANA702163 MANA702164 MANA702165 MANA702402
Lokk, Skrulokk for skruhals hetteglass, N 9
Lokk Lokk til vials
The suitable screw closures are ready assembled.
  • Large range of screw closures with different septum materials and cap colours
  • Including a magnetic version for CTC GC PAL autosampler or identical constructions
  • Bonded screw caps (septa firmly connected with the cap) have the advantage that blunt HPLC needles cannot push the septum into the vial
  • Septa made from economical Red Rubber/FEP, high quality silicone/PTFE (also slitted) or silicone/polyimide and from virgin PTFE