Size exclusion chromatography columns, ProSEC 300S
Leverandør: VARIAN
PL1147-1501 PL1147-6501 PL1547-1501 PL1547-5501
VARIPL1147-1501EA 18575 NOK
VARIPL1147-1501 VARIPL1147-6501 VARIPL1547-1501 VARIPL1547-5501
Size exclusion chromatography columns, ProSEC 300S
Kromatografikolonner
The ProSEC 300S column is specifically designed as a single column solution for globular protein analysis. The pore size selection and optimisation provides an extended linear resolving range so that this single column can be used for analysis across the full range of globular proteins.
  • Mechanically robust polymer particles that do not bleed during use
  • Single column with extended linear resolving range
  • Column dimensions for use with multi-detector systems
This gives exceptionally stable baselines making this column an ideal choice for use with lightMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks. trykk 250 bar
pH område 2 - 7,5