Sentino® Dispenser Pack Refills for Membrane Filter Dispenser
Leverandør: Cytiva (Formerly Pall Lab)
68121 68123 68124 68125 68121ME
514-0628EA 8550 NOK
514-0628 514-0629 514-0630 514-0631 PALL68121ME
Sentino® Dispenser Pack Refills for Membrane Filter Dispenser
Filter Membranfilter
These membranes have been designed specifically for use with the Sentino® automatic filter dispenser.
  • Ideal for high volume laboratories
  • Eliminate the time and waste of individually sealed packs of membrane
Leveringsinformasjon: Each box contains two additional discs for loading the dispenser.
Bestill nå
SPESIFIKASJONER
Type GN-6 Metricel® Supor® Metricel® Black Metricel® Black ME25
Environmentally Preferable
Porestørrelse 0,45 µm 0,20 µm 0,45 µm 0,80 µm 0,45 µm
Tykkelse 152 µm 145 µm 117 µm 135 µm _
Vanngjennomstrømning ≥65,0 ml/min/cm² (0,7 bar) ≥19,3 ml/min/cm² (0,7 bar) ≥34,5 ml/min/cm² (0,7 bar) ≥102,8 ml/min/cm² (0,7 bar) _
Materiale Cellulose mixed esters Polyethersulfone (PES) Polyethersulfone (PES) Polyethersulfone (PES) Cellulose mixed esters