Spectrocolorimeter, LC 100, Lovibond®
Supplier: Lovibond Tintometer
401100 401120
633-0242EA 33075 NOK
633-0242 633-0243
Spectrocolorimeter, LC 100, Lovibond®
Colorimeters Reflectance Colorimeters
A lightweight, handheld imaging spectrocolorimeter that enables setting and maintaining colour standards throughout the manufacturing process. In less than 2 seconds the instrument provides, not only a reading, but also a report telling how close the colour is to a measured standard. The instrument takes images of the sample using 8 different visible illuminations and 1 ultraviolet LED (9 bands in total) for more accurate colour measurement. Each measurement is a combination of 27 images illuminated with different colours from different directions.
  • Measurement of food, powders
More Product Information
Order Now
SPECIFICATIONS
Model LC 100
Environmentally Preferable
Illuminants D65
Light source 25 independent tridirectional LEDs
Measuring area 4 + 8 mm
Observer A (10)
Measuring geometry 45/0 image capture
Reproducibility short term typically 0,10 ΔE 94 on white (d65/10)
Measurement time 1,8 s
Display 45 mm colour TFT
Storable data sets 20 standards/350 samples
Interfaces USB