3-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid

E0848-1G E0848-5G
TCIAE0848-1GEA 300 NOK
TCIAE0848-1G TCIAE0848-5G
3-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Ethoxycarbonyl)phenylboronic acid
Formel: C₉H₁₁BO₄
MW: 193,99 g/mol
Smeltepunkt: 138°C
MDL nummer: MFCD02179444
CAS-nummer: 4334-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eda939938aa4a56ec21c32887a69385bb2f13809&vid=eda939938aa4a56ec21c32887a69385bb2f13809&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR