2-Amino-6-chloro-4-nitrophenol ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Chloro-2-hydroxy-5-nitroaniline
A2245-25G
A2245-5G
TCIAA2245-25GEA
2950
NOK
TCIAA2245-25G
TCIAA2245-5G
2-Amino-6-chloro-4-nitrophenol ≥98.0% (by titrimetric analysis)
2-Amino-6-chloro-4-nitrophenol
Formel:
C₆H₅ClN₂O₃ MW: 188,57 g/mol |
MDL nummer:
MFCD00035767 CAS-nummer: 6358-09-4 EINECS: 228-762-1 |
Spesifikasjon Testresultater
Appearance | Orange to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...