Retinol all trans (synthetic) 95%
Leverandør: Thermo Scientific
|
Danger
|
J62079.03EA
4925
NOK
J62079.03
Retinol all trans (synthetic) 95%
Retinol all trans
Formel:
C₂₀H₃₀O MW: 286,46 g/mol Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD00001552 CAS-nummer: 68-26-8 EINECS: 200-683-7 Merck Indeks: 14,10013 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...