(R)-(-)-2-Chloromandelic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: (R)-2-(2-Chlorophenyl)-2-hydroxyacetic acid
C1663-1G C1663-5G
TCIAC1663-1GEA 230 NOK
TCIAC1663-1G TCIAC1663-5G
(R)-(-)-2-Chloromandelic acid ≥98.0% (by GC, titration analysis)
(R)-(-)-2-Chloromandelic acid
Formel: C₈H₇ClO₃
MW: 186,59 g/mol
MDL nummer: MFCD00798429
CAS-nummer: 52950-18-2
EINECS: 444-040-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6181db597d7d0dace190df4c387371a0bc828a93&vid=6181db597d7d0dace190df4c387371a0bc828a93&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C