VWR® Dehydrert kulturmedier, pulverisert
Leverandør: VWR Chemicals
84646.0500EA 2625 NOK
84646.0500 84647.0500 84648.0500 84649.0500 84651.0500 84652.0500 84653.0500 84641.0500 84643.0500 84644.0500 84645.0500 84654.0500 84655.0500 84656.0500 84657.0500 84658.0500 84659.0500 84660.0500 84661.0500 84662.0500 84663.0500 84664.0500 84665.0500 84666.0500 84667.0500 84668.0500 84669.0500 84670.0500 84671.0500 84673.0500 84748.0500 84684.0500 84685.0500 84686.0500 84674.0500 84678.0500 84679.0500 84680.0500 84681.0500 84682.0500 84683.0500 84689.0500 84690.0500 84691.0500 84694.0500 84631.0500 84600.0500 84617.0500 84619.0500 84621.0500 84749.0500 84630.0500 84614.0500 84624.0500 84625.0500 84626.0500 84627.0500 84629.0500 84622.0500 84602.0500 84603.0500 84604.0500 84605.0500 84606.0500 84608.0500 84632.0500 84611.0500 84612.0500 84613.0500 84633.0500 84607.0500 84695.0500 84634.0500 84635.0500 84636.0500 84637.0500 84638.0500 84650.0500 84696.0500 84697.0500 84698.0500 84699.0500 84642.0500 84639.0500 84675.0500 84677.0500 84855.0500 84855.5000 84600.5000 84901.0500 84674.5000 84600.25000 84611.5000 84684.5000 84649.5000 84902.0500 84905.0500 84906.0500 84907.0500 84903.0500 84909.0500 84675.5000 84677.5000 84681.5000 84689.5000 84950.0500 84608.5000 84621.5000 84951.5000 84900.0500 85050.0500 85051.0500 85052.0500 84629.5000 85054.0500 84954.5000 84679.5000 85055.0500 84956.0500 85056.0500 84957.0500 85940.0500 85942.0500 84626.5000 85945.0500 85943.0500 85946.0500 85947.5000 85837.0500 84683.5000 84695.5000 85838.0500 85839.0500 84646.5000 84951.20000 84602.5000 85863.0500 84675.25000 84679.25000 84603.5000 85877.0500 84677.10000 84678.5000 85881.0500 85887.0500 85887.5000 85883.0500 85885.0500 85885.5000 85888.0500 85888.5000 85889.0500 85889.5000 85890.0500 85891.0500 84951.0500 85894.5000 92427.0500
VWR® Dehydrert kulturmedier, pulverisert
Mikrobiologiske medier Media, dehydrert for mikrobiologi
A range of culture media in compliance with ISO standards and the International Harmonised Pharmacopoeia. All our media are, as you would expect, produced using high quality raw materials and are subject to strict quality controls, in a production environment following Good Manufacturing Practice guidelines. Raw materials are selected in compliance with the required standards. Dehydrated culture media should be stored at 10 to 30 °C unless otherwise stated, in their closed containers in a dry environment. Standard pack size is 500 g.
  • Compliant with international standards, such as ISO and Pharmacopoeia
  • Media growth promotion testing performed is compliance with Pharmacopoeias and ISO 11133
  • Available in animal-free standard formulations
  • Custom products available