Cation exchange chromatography columns, HiTrap™ SP XL
Leverandør: Cytiva
17-5160-01 17-5161-01
17-5160-01EA 1925 NOK
17-5160-01 17-5161-01
Cation exchange chromatography columns, HiTrap™ SP XL
Kromatografikolonner
HiTrap SP XL are strong cation exchangers prepacked with SP Sepharose XL media, optimized for fast, convenient small-scale protein capture using ion exchange (IEX) chromatography.
  • High binding capacities with high flow rates
  • Convenient and affordable for fast, easy separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump, or chromatography system such as ÄKTA design
Q Sepharose XL and SP Sepharose XL resins have long chains of dextran coupled to a robust, 6% highly cross-linked agarose matrix. The dextran chainsMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks. trykk 5 bar
pH område 4 - 13