5-Formyl-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 5-Formyl-2-methoxybenzeneboronic acid, (5-Formyl-2-methoxyphenyl)boronic acid

F0834-1G F0834-5G
TCIAF0834-1GEA 405 NOK
TCIAF0834-1G TCIAF0834-5G
5-Formyl-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
5-Formyl-2-methoxyphenylboronic acid
Formel: C₈H₉BO₄
MW: 179,9700 g/mol
MDL nummer: MFCD01319044
CAS-nummer: 127972-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8687f944f9a2ab8f6ebc7a5b080dcae4ae8a76d&vid=c8687f944f9a2ab8f6ebc7a5b080dcae4ae8a76d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR