2-Chloro-4'-fluorobenzophenone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (2-Chlorophenyl)(4-fluorophenyl)methanone

C1860-25G C1860-5G
TCIAC1860-25GEA 2350 NOK
TCIAC1860-25G TCIAC1860-5G
2-Chloro-4'-fluorobenzophenone ≥98.0% (by GC)
2-Chloro-4'-fluorobenzophenone
Formel: C₁₃H₈ClFO
MW: 234,6534 g/mol
MDL nummer: MFCD00000559
CAS-nummer: 1806-23-1
EINECS: 217-300-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f78ee328be2036e18ed6371a8b93ed3f74beda9&vid=1f78ee328be2036e18ed6371a8b93ed3f74beda9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR