4-(1-Pyrrolidinyl)pyridine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
4-Pyrrolidinopyridine
P0939-25G
P0939-5G
TCIAP0939-25GEA
1300
NOK
TCIAP0939-25G
TCIAP0939-5G
4-(1-Pyrrolidinyl)pyridine ≥98.0% (by titrimetric analysis)
4-(1-Pyrrolidinyl)pyridine
Formel:
C₉H₁₂N₂ MW: 148,21 g/mol Kokepunkt: 170°C |
MDL nummer:
MFCD00006415 CAS-nummer: 2456-81-7 EINECS: 219-534-2 FN: 2923 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 94.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 56.0 to 61.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...