1,3,5-Triethynylbenzene ≥98.0% (by GC)

Leverandør: TCI

T2760-1G T2760-5G
TCIAT2760-1GEA 1625 NOK
TCIAT2760-1G TCIAT2760-5G
1,3,5-Triethynylbenzene ≥98.0% (by GC)
1,3,5-Triethynylbenzene
Formel: C₁₂H₆
MW: 150,18 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00068621
CAS-nummer: 7567-63-7
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fa68528bbb84644c322fe8380eb6efadedf3d8e&vid=4fa68528bbb84644c322fe8380eb6efadedf3d8e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR