4-Amino-1-boc-piperidine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester

A2045-1G A2045-5G
TCIAA2045-1GEA 420 NOK
TCIAA2045-1G TCIAA2045-5G
4-Amino-1-boc-piperidine ≥97.0% (by GC, titration analysis)
4-Amino-1-Boc-piperidine
Formel: C₁₀H₂₀N₂O₂
MW: 200,28 g/mol
MDL nummer: MFCD01076201
CAS-nummer: 87120-72-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c09add56dea5019957382efdcea454af528872d&vid=6c09add56dea5019957382efdcea454af528872d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 43.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR