(S)-(+)-2,2-Dimethylcyclopropanecarboxamide ≥98.0% (by GC)

Leverandør: TCI

Synonymer: (S)-2,2-Dimethylcyclopropanecarboxamide

D3676-25G D3676-5G
TCIAD3676-25GEA 1150 NOK
TCIAD3676-25G TCIAD3676-5G
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide ≥98.0% (by GC)
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide
Formel: C₆H₁₁NO
MW: 113.1600 g/mol
MDL nummer: MFCD00216614
CAS-nummer: 75885-58-4
EINECS: 278-334-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6ea062e9ac3f62d86447d9e68f8786d9d44d74c&vid=c6ea062e9ac3f62d86447d9e68f8786d9d44d74c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 135.0 to 140.0 deg-C
Specific rotation [a]20/D +78.0 to +88.0 deg (C=1, methanol)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR