trans,trans-Dibenzylideneacetone ≥99.0% (by GC)
Leverandør: TCI
Synonymer:
(1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one
D0903-25G
D0903-250G
TCIAD0903-25GEA
650
NOK
TCIAD0903-25G
TCIAD0903-250G
trans,trans-Dibenzylideneacetone ≥99.0% (by GC)
trans,trans-Dibenzylideneacetone
Formel:
C₁₇H₁₄O MW: 234,30 g/mol |
MDL nummer:
MFCD00004790 CAS-nummer: 35225-79-7 EINECS: 208-697-5 Merck Indeks: 14,03006 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 111.0 to 114.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...