2-Chloro-5-nitrobenzophenone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (2-Chloro-5-nitrophenyl)(phenyl)methanone

C1776-25G C1776-5G
TCIAC1776-25GEA 475 NOK
TCIAC1776-25G TCIAC1776-5G
2-Chloro-5-nitrobenzophenone ≥98.0% (by GC)
2-Chloro-5-nitrobenzophenone
Formel: C₁₃H₈ClNO₃
MW: 261,66 g/mol
MDL nummer: MFCD00007295
CAS-nummer: 34052-37-4
EINECS: 251-811-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5263d288d19ad4824e3ec519aef22bf3ccd3d56f&vid=5263d288d19ad4824e3ec519aef22bf3ccd3d56f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR