2-Hydroxy-4-methoxy-5-sulphobenzophenone hydrate ≥98.0% (by titrimetric analysis) (contains 5-10% Isopropyl Alcohol)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Hydroxy-4-methoxybenzophenone-5-sulphonic Acid Hydrate
H0466-25G
H0466-500G
TCIAH0466-25GEA
555
NOK
TCIAH0466-25G
TCIAH0466-500G
2-Hydroxy-4-methoxy-5-sulphobenzophenone hydrate ≥98.0% (by titrimetric analysis) (contains 5-10% Isopropyl Alcohol)
2-Hydroxy-4-methoxy-5-sulphobenzophenone hydrate
Formel:
C₁₄H₁₂O₆S·H₂O MW: 308,31 g/mol (anhy) |
MDL nummer:
MFCD00024962 CAS-nummer: 4065-45-6 EINECS: 223-772-2 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 98.0 % (calcd.on dried substance) |
Drying loss | max. 22.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...