5'-Chloro-2'-hydroxy-4'-methylacetophenone ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-(5-Chloro-2-hydroxy-4-methylphenyl)ethanone

C1629-25G C1629-5G
TCIAC1629-25GEA 620 NOK
TCIAC1629-25G TCIAC1629-5G
5'-Chloro-2'-hydroxy-4'-methylacetophenone ≥95.0% (by GC)
5'-Chloro-2'-hydroxy-4'-methylacetophenone
Formel: C₉H₉ClO₂
MW: 184,6200 g/mol
Kokepunkt: 137°C
MDL nummer: MFCD00191920
CAS-nummer: 28480-70-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cb4f8116ffc2f9737854213c18c2155a4a97ac4&vid=7cb4f8116ffc2f9737854213c18c2155a4a97ac4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR