Isoflurane ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether

C2485-25G C2485-5G
TCIAC2485-25GEA 1200 NOK
TCIAC2485-25G TCIAC2485-5G
Isoflurane ≥98.0% (by GC)
Isoflurane
Formel: C₃H₂ClF₅O
MW: 184,49 g/mol
Kokepunkt: 49°C
MDL nummer: MFCD00066609
CAS-nummer: 26675-46-7
EINECS: 247-897-7
Merck Indeks: 14,05175
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b96833421ef1a4ec4f5bd47b8cdc4296e37bb97&vid=3b96833421ef1a4ec4f5bd47b8cdc4296e37bb97&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR