2-Methyl-3-hexanol

Leverandør: TCI
Warning

M1266-25ML M1266-5ML
TCIAM1266-25MLEA 1300 NOK
TCIAM1266-25ML TCIAM1266-5ML
2-Methyl-3-hexanol
2-Methyl-3-hexanol
Formel: C₇H₁₆O
MW: 116,201 g/mol
Kokepunkt: 145°C
Flash punkt: 41°C
MDL nummer: MFCD00004577
CAS-nummer: 617-29-8
EINECS: 210-508-6
FN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3fc30c35ae25c4200e5509e9ba1e45773e3fd38&vid=d3fc30c35ae25c4200e5509e9ba1e45773e3fd38&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR