D-(+)-Turanose ≥98.0% (by HPLC)
Leverandør: TCI
Synonymer:
(3S,4R,5R)-1,4,5,6-Tetrahydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-one, Turanose
T0542-1G
T0542-5G
TCIAT0542-1GEA
555
NOK
TCIAT0542-1G
TCIAT0542-5G
D-(+)-Turanose ≥98.0% (by HPLC)
D-(+)-Turanose
Formel:
C₁₂H₂₂O₁₁ MW: 342,30 g/mol |
MDL nummer:
MFCD00006606 CAS-nummer: 547-25-1 EINECS: 208-918-5 Merck Indeks: 14,09821 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Specific rotation [a]20/D | +74.0 to +77.0 deg (C=4, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...