4-Amino-2,5-xylenol ≥97.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Amino-2,5-dimethylphenol
A2116-25G
A2116-5G
TCIAA2116-25GEA
4175
NOK
TCIAA2116-25G
TCIAA2116-5G
4-Amino-2,5-xylenol ≥97.0% (by GC, titration analysis)
4-Amino-2,5-xylenol
Formel:
C₈H₁₁NO MW: 137,1800 g/mol Smeltepunkt: 238°C |
MDL nummer:
MFCD00007873 CAS-nummer: 3096-71-7 EINECS: 221-449-0 |
Spesifikasjon Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...