4-Chloro-o-toluic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
C2508-5G C2508-25G
TCIAC2508-5GEA 915 NOK
TCIAC2508-5G TCIAC2508-25G
4-Chloro-o-toluic acid ≥98.0% (by GC, titration analysis)
4-Chloro-o-toluic acid
Formel: C₈H₇ClO₂
MW: 170,6 g/mol
MDL nummer: MFCD00045852
CAS-nummer: 7499-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d3060dcf804ec273d9fc342ea2d0f7eb271bdc7&vid=8d3060dcf804ec273d9fc342ea2d0f7eb271bdc7&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 171.0 to 175.0 deg-C