Propyltriethoxysilane ≥98.0% (by GC)
Leverandør: TCI
T2867-25G
T2867-100G
TCIAT2867-25GEA
340
NOK
TCIAT2867-25G
TCIAT2867-100G
Propyltriethoxysilane ≥98.0% (by GC)
Propyltriethoxysilane
Formel:
C₉H₂₂O₃Si MW: 206,36 g/mol |
MDL nummer:
MFCD00026756 CAS-nummer: 2550-02-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...