D(-)-Lysine monohydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: (R)-(-)-Lysine monohydrochloride, H-D-Lys-OH.HCl, D-Lysine hydrochloride, (R)-(-)-2,6-Diaminocaproic acid monohydrochloride, (R)-(-)-2,6-Diaminohexanoic acid monohydrochloride

L0128-25G L0128-5G
TCIAL0128-25GEA 2675 NOK
TCIAL0128-25G TCIAL0128-5G
D(-)-Lysine monohydrochloride ≥98.0% (by titrimetric analysis)
D(-)-Lysine monohydrochloride
Formel: C₆H₁₅ClN₂O₂
MW: 182,65 g/mol
Smeltepunkt: 261…262 °C
MDL nummer: MFCD00012920
CAS-nummer: 7274-88-6
EINECS: 230-691-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eef64c80bb80f86ebd8de70f6267fb42cfb3ffca&vid=eef64c80bb80f86ebd8de70f6267fb42cfb3ffca&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -20.0 to -24.0 deg (C=8, HCl (1+1))

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR