3,5-Dibromoanthranilic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 2-Amino-3,5-dibromobenzoicacid, 2-Amino-3,5-dibromobenzoic acid
D4177-5G D4177-25G
TCIAD4177-5GEA 395 NOK
TCIAD4177-5G TCIAD4177-25G
3,5-Dibromoanthranilic acid ≥98.0% (by HPLC, titration analysis)
3,5-Dibromoanthranilic acid
Formel: C₇H₅Br₂NO₂
MW: 294,93 g/mol
MDL nummer: MFCD00017087
CAS-nummer: 609-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd00f1156a04cc0fc379bbc7e5e92697e323d72d&vid=b10659d9b450bed8617583290394da4b8a4c09b3&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 234.0 to 238.0 deg-C