4-[(Trifluoromethyl)thio]aniline ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-((Trifluoromethyl)thio)aniline

T2775-25G T2775-5G
TCIAT2775-25GEA 2775 NOK
TCIAT2775-25G TCIAT2775-5G
4-[(Trifluoromethyl)thio]aniline ≥98.0% (by GC, titration analysis)
4-[(Trifluoromethyl)thio]aniline
Formel: C₇H₆F₃NS
MW: 193.19 g/mol
Flash punkt: 107°C
MDL nummer: MFCD00040926
CAS-nummer: 372-16-7
EINECS: 206-744-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8114b7a8c140822201d12d5b42c83da45e4f95c5&vid=8114b7a8c140822201d12d5b42c83da45e4f95c5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR