Container, wide mouth, with screw cap, Azlon®
Leverandør: SciLabware
JWP0030P JWP0060P JWP0125P JWP0250P JWP0500P JWP0950P JWP1200P
216-1352EA 1375 NOK
216-1352 216-1354 216-1355 216-1356 216-1357 216-1380 216-1360
Container, wide mouth, with screw cap, Azlon®
Prøvecontainere Beholdere, generelt bruk
PP, translucent.
  • Autoclavable to 121 °C
  • Good chemical resistance
  • With unlined screw caps
Ideal for storage of samples, powders, animal feeds etc.