HPLC columns, Hypersil GOLD™ Phenyl
Leverandør: Thermo Fisher Scientific
25903-154630 25903-102130 25902-102130 25902-052130 25905-154630 25903-104630 25905-012101 25905-254630 25905-102130 25903-014001 25903-013001 25903-012101 25902-152130
554-4495EA 12150 NOK
554-4495 555-5147 555-5748 554-4323 555-5126 555-5150 555-5156 554-4325 555-5116 555-5755 555-5754 555-5753 555-5750
HPLC columns, Hypersil GOLD™ Phenyl
Kromatografikolonner
Contains a C4 linker which allows for superior alignment of the phenyl ring with aromatic molecules. Get excellent retention and unique selectivity for aromatic and moderately polar analytes with Thermo Scientific™ Hypersil GOLD™ Phenyl reversed phase HPLC columns.
  • Enhancing pi-pi interactions and improving the retention of the molecules
  • Moderate hydrophobicity
  • Outstanding peak shape results in greater sensitivity
A butyl (C4) linker in the phenyl bonded phase allows for superior alignment of the phenyl ring with aromatic molecules, thus enhancing pi-More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 8,50% 8.50%
pH område 2 to 8 2 to 8
Overflate areal 220 m²/g 220 m²/g