HPLC columns, Hypersil™ BDS C18
Leverandør: Thermo Fisher Scientific
28105-254630 28105-104630 28105-103030 28105-102130 28105-054630 28105-052130 28105-014001 28105-012101 28105-124030 28103-154630 28103-153030 28103-152130 28103-104630 28103-104030 28103-103030 28105-013001 28105-253030 28105-123030 28105-204630 28105-154630 28105-154030 28105-153030 28105-152130 28105-124630 28103-053030 28105-254030 28103-102130 28103-052130 28103-014001 28103-012101 28102-154630 28102-104630 28102-054630 28103-054630 28102-152130 28103-054030 28103-033030 28103-032130 28105-053030 28102-102130 28102-052130 28102-034630 28102-014001 28103-013001 28103-154030 28105-104030 28105-012106 28105-304030 28105-304630
554-5310EA 11125 NOK
554-5310 555-6667 555-6665 555-6664 555-6662 555-6659 554-5312 555-6596 555-6670 555-6588 555-6586 555-6585 554-5309 555-6579 555-6578 555-6651 555-6626 555-6669 555-6624 555-6675 555-6674 555-6673 555-6672 555-6671 555-6572 555-6627 555-6577 555-6571 554-5311 555-6612 555-6609 555-6607 555-6605 555-6574 555-6608 555-6573 555-6620 555-6619 555-6660 555-6606 555-6603 555-6602 555-6600 555-6614 555-6587 555-6666 555-6597 555-6630 555-6631
HPLC columns, Hypersil™ BDS C18
Kromatografikolonner
The Hypersil™ BDS C18 with the base deactivated silica backbone has reduced silanol activity and hence produce reliable results for QA/QC labs.
  • Base deactivated with minimal residual silanol activity
  • Good reproducibility
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 11% 11% 11% 11% 11% 11% 11% 11%
Kappet ved endene Ja Ja Ja Ja Ja Ja Ja Ja
Overflate areal 170 m²/g 170 m²/g 170 m²/g 170 m²/g 170 m²/g 170 m²/g 170 m²/g 170 m²/g
Temperaturgrense 60 °C 60 °C 60 °C 60 °C 60 °C 60 °C 60 °C 60 °C
USP kode L1 L1 L1 L1 L1 L1 L1 L1