(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Leverandør: Novabiochem (Part of Merck)
Warning
Synonymer: PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate
8.51009.0100 8.51009.0025
01-62-0016-100EA 0 NOK
01-62-0016-100 01-62-0016-25
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Formel: C₁₈H₂₈F₆N₆OP₂
MW: 520,4 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00077411
CAS-nummer: 128625-52-5