(±)-2-Piperazinecarboxylic acid dihydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Piperazine-2-carboxylic acid dihydrochloride

P1391-25G P1391-5G
TCIAP1391-25GEA 1775 NOK
TCIAP1391-25G TCIAP1391-5G
(±)-2-Piperazinecarboxylic acid dihydrochloride ≥98.0% (by titrimetric analysis)
(±)-2-Piperazinecarboxylic acid dihydrochloride
Formel: C₅H₁₂Cl₂N₂O₂
MW: 203,07 g/mol
MDL nummer: MFCD00044803
CAS-nummer: 3022-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9602076237fc9a7c86a3c958a5567fbc4889e8d6&vid=9602076237fc9a7c86a3c958a5567fbc4889e8d6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR