1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulphonate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,1,1,3,3,3-Hexafluoropropan-2-yl 4-methylbenzenesulphonate

H1281-25G H1281-5G
TCIAH1281-25GEA 2200 NOK
TCIAH1281-25G TCIAH1281-5G
1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulphonate ≥98.0% (by GC)
1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulphonate
Formel: C₁₀H₈F₆O₃S
MW: 322,23 g/mol
Kokepunkt: 134°C
MDL nummer: MFCD00039262
CAS-nummer: 67674-48-0
EINECS: 266-887-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e141aba057a29a1c6f027c4c6747d589e050aa85&vid=e141aba057a29a1c6f027c4c6747d589e050aa85&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR